- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 594
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western Blot:Yes
- Size:1.0 mg
- Cross Adsorption:Yes
- Format:Purified IgG
- Form:Lyophilized
- Storage Buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
- Storage Temperature:2-8 °C
- Concentration:1 mg/ml (E 1% at 280 nm = 13.0)
- Shipping Temperature:2-8 °C
- Immunogen:Rabbit IgG
- Tested Applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phaseRabbit IgG
- Cat. No.:10149-296
- Supplier no.:IR1219
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593nm / 618nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit