Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (DyLight® 594)
Anti-IgG Donkey Polyclonal Antibody (DyLight® 594)
  10149-222
undefined
Anti-IgG Donkey Polyclonal Antibody (DyLight® 594)
  10149-222
 :  DKXMU-003-F594NHSX

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 594
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Cross Adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Mouse IgG
  • Tested Applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseMouse IgG
  • Cat. No.:
    10149-222
  • Supplier no.:
    IR1170

 

 

The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593nm / 618nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse