Molecular sieve 3A (0.3 nm, 3 Å), Supelco®
EM1.05734.1000
:
- Pack type:Plastic bottle for solids
- Size:1 Kg
- Cat. No.:EM1.05734.1000
- Supplier no.:1.05734.1000
:
:M-Clarity™ quality level = MQ200
:308080-99-1
:1318-02-1
:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
:MFCD00131613