

2-Propanol, Prepsolv® for preparative chromatography, Supelco®
EM1.13350.2500
:
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM1.13350.2500
- Supplier no.:1.13350.2500
:
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,
:M-Clarity™ quality level = MQ200
:01-2119457558-25
:GHS02
:GHS07
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674