



2,2,4-Trimethylpentane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–12 °C
:GHS02
:GHS08
:GHS07
:GHS09