

2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (lit.)
Melting Pt:–89.5 °C (lit.)
Density:0.785 g/ml (25°C) (lit.)
EINECS:200-661-7
UN:1219
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:12 °C
REACH:01-2119457558-25
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together