


1-Butanol, EMPLURA®, Supelco®
EM8.22262.2500
:
- Pack type:Plastic bottle
- Size:2.5 L
- Cat. No.:EM8.22262.2500
- Supplier no.:8.22262.2500
:
:74.12 g/mol
:13,01539
:117.6 °C (1013 hPa)
:117.6 °C (1013 hPa)
:–89 °C
:-89 °C
:0.812 g/cm³ (20 °C)
:30 °C
:30 °C
:Ambient
:200-751-6
:1120
:3,III
:M-Clarity™ quality level = MQ200
:01-2119484630-38
:GHS02
:GHS05
:GHS07
:71-36-3
:H₃C(CH₂)₂CH₂OH
:MFCD00002964