4-Blade PTFE Agitators, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
The 4-Blade PTFE agitator is available with either a 45° or 90° blade angle.
- Designed for use with glass or metal shafts
- Secured with a compression-style fitting allowing it to be positioned anywhere along the length of the shaft
Related Information
VF1012 10 to 20L Jackted Reactors VF1016 20L and 50L Ready Reactors