Microplate Self-adhesive Film and Foil
Supplier: Eppendorf
Certificates
About this item
Secure your samples with Eppendorf® Microplate self-adhesive film and foil, perfect for incubation and short-term storage. Films protect from punctures while aluminum foils shield light-sensitive samples with easy sealing.
- Storage films are transparent and provide protection against unwanted punctures
- Storage foils are made of aluminum, can be pierced, and protect light-sensitive samples
- Storage film and foil glued seals for simple and dependable sealing during sample storage
For long-term storage, we recommend the use of an Eppendorf heat sealing film.