ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics
Gold Standard Diagnostics offers a comprehensive menu of ELISA assays for Infectious Disease and Autoimmune diagnostic testing
Most kits feature interchangeable reagent components, consistent protocols and break-away wells.
Our comprehensive menu of assays fits the needs of virtually any laboratory.
: FDA Cleared or Exempt.
: Package contains microtiter plate(s), dilution buffer, wash buffer, controls, conjuage and stop solution.
- Primary antibody reactivity:Human
- Target protein:Free Thyroxine (fT4)
- Sample Type:Serum
- Application:Endocrinology
- Assay Type (ELISA with LOV):Competitive