Advantec Pressure Vessels are used to contain process fluids for filtration through pressure type filter holders
Use under positive pressure or negative pressure. When working with negative pressure or operating at low pressure(<30psi), use the replacement vacuum closure.
Type 316/316L stainless steel complies with good manufacturing practices and is recommended for low pH and high salt applications. Type 304 stainless steel Standard Closure vessels offer 155psi (1–5 gallons) or 115psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 304 stainless steel Wide Mouth Closure vessels offer 125psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 316L stainless steel Standard Closure vessels offer 150psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F). Type 316L stainless steel Wide Mouth Closure vessels offer 125psi (1–5 gallons) or 110psi (8–16 gallons) maximum operating pressure at 149°C (300°F).
The pressure vessels are available in type 304 or 316L stainless steel. Choose either standard closure or wide mouth closure (for ease of cleaning). Electropolished finish provides a smooth surface for cleaning. Molded rubber skirt/base protects vessel against rough handling (also available with stainless base for continuous operation at high temperatures).
- Catalog No:
- 101105-884
- 101105-850
- 101105-822
- 101105-804
- 101105-764
- 101105-800
- 101105-650
- 101105-246
- 101105-796
- 101105-872
- 101105-798
- 101105-840
- 101105-826
- 101105-890
- 101105-834
- 101105-882
- 101105-888
- 101105-892
- 101105-838
- 101105-828
- 101105-750
- 101105-830
- 101105-832
- Closure Size:
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Standard: 3.875 x 3.25", Wide Mouth: 5.875 x 4.875"
- Inlet/Outlet Connections:
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- 4 Ports: 0.25" NPTF
- Skirt:
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel
- 1–5 gal. Vessels: Neoprene Rubber, 8–16 gal. Vessels: Stainless Steel