Orders LinkContactUsLinkComponent
Advansta Blot Development Folders
Advansta Blot Development Folders
Supplier:   Advansta
Advansta Blot Development Folders
Supplier:  Advansta
Certificates
Advansta Blot Development Folders
Advansta Blot Development Folders
2 Options Available Below
Product Details & Documents
Advansta's blot development folders are transparent plastic folders that hold your blot while it is exposed to film or a CCD camera. These folders keep your blot flat for perfect film exposures, avoiding the wrinkling or bunching that can occur with plastic wrap.
  • Clear images
  • Suitable for all blots
  • Safe blot handling
  • Convenient size
Development folders are suitable for chemiluminescence and fluorescence imaging. You can move your blot without scratching of putting pressure on the surface. Folders allow you to image up to two blots imultaneously in one folder.
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • 490016-806
    Blot Development Folders
    Blot Development Folders
    5 x 7"
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $119.24
    $119.24
    Pack of 25
       $119.24
    Minimum order quantity is 1
    Supplier Number: L-07020-025
  • 490016-808
    Blot Development Folders
    Blot Development Folders
    5 x 7"
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $231.65
    $231.65
    Pack of 100
       $231.65
    Minimum order quantity is 1
    Supplier Number: L-07020-100