C-Leg Stand-Alone Corner Laboratory Benches, Production Basics
Supplier: Production Basics
Certificates
About this item
Use this free-standing workstation to create corner or wrap-around configurations with C-Leg Laboratory Benches or Add-ons.
- Finished with rounded corners and protective T-mold edge
Compatible with standard above-worksurface accessories.
Accessories information: For use with on any C-Leg Lab Bench. Add uprights to mount accessories on RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench, Basic Organizer or Easy Lift 4-post Lab Bench.
Ordering information: Specify a Frame and Laminate color otherwise will be default Blue Frame/White Laminate. Frame available in Light Neutral/Almond, Blue, White, Grey and Black. Worksurface available in White, Grey and Beige. The two (2) included 162.6 cm (64") high integrated back frames are ready to accept accessories. Accessories are sold separately. Order same width accessory as workstation width.
Delivery: Includes (2) single-sided frame, (2) C-leg, adjustable, (1) corner worksurface, (1) support rail, (8) leveling feet, (1) parts and hardware packet.