Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 104]
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 104]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 104]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-506
  • 10051-504
  • Antigen Symbol:
  • PTPRC
  • PTPRC
  • Antigen Name:
  • Protein Tyrosine Phosphatase Receptor Type C
  • Protein Tyrosine Phosphatase Receptor Type C
  • Antigen Synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • 104
  • 104
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information