Rat IgG2a Monoclonal Isotype Control [clone: 2A3] (PE (Phycoerythrin))
10051-330
- Antibody Type:Control
- Antigen Name:UDP-glucuronosyltransferase 2A3
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10051-330
- Supplier no.:50-4321-U100
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: 2A3
Clonality: Monoclonal
Clone: 2A3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: