Orders LinkContactUsLinkComponent
VWR® Basic Frosted Microscope Slide without Tissue Paper
VWR® Basic Frosted Microscope Slide without Tissue Paper
Slides, 1.0 mm, 75×25 mm (3×1")
Catalog # 89085-399
Supplier:   VWR International
VWR® Basic Frosted Microscope Slide without Tissue Paper
Catalog # 89085-399
Supplier:  VWR International
VWR® Basic Frosted Microscope Slide without Tissue Paper
VWR® Basic Frosted Microscope Slide without Tissue Paper
Slides, 1.0 mm, 75×25 mm (3×1")
Price
$453.86
Log in for availability
Case of 1,440
Product Details & Documents
VWR® Frosted Microscope Slides without Tissue Paper are specially designed with consistent and clean surfaces for staining experiments.
  • Consistent, clean surfaces
  • Frosted edge for marking
They are ideal for histology, hematology, and microbiology applications. Slides feature frosted areas on both sides of one end for convenient marking. A fine polished edge and 45° clipped corners promote ease and safety in handling. Slides are packaged 72 per pack, 20 packs per case.
Specifications
  • Description:
    Slides
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm