Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Storage Rack for Microscope Slide Boxes

Storage Rack for Microscope Slide Boxes

Storage Rack for Microscope Slide Boxes, Clear, 711×171×228 mm
Catalog # 490007-570
Supplier:   Heathrow Scientific
Supplier #:  HS159890B
Storage Rack for Microscope Slide Boxes
Catalog # 490007-570
Storage Rack for Microscope Slide Boxes
Storage Rack for Microscope Slide Boxes

Storage Rack for Microscope Slide Boxes, Clear, 711×171×228 mm

Price
$650.87
Log in for availability
Each
Product Details & Documents

Storage racks simplify organization for easy sample sorting and selection.

  • The clear acrylic construction allows you to see the labels on the slide boxes
  • Place boxes in rows or columns by color to create an easy coding system

Each rack holds up to 15 slide boxes.

Specifications
  • Color:
    Clear
  • Description:
    Storage Rack for Microscope Slide Boxes
  • Dimensions:
    711×171×228 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.