ContactUsLinkComponent
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
Catalog # 10147-000
undefined
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
Catalog # 10147-000
Supplier Number:  376814
Specifications
  • Sterility:
    Sterile
  • Volume:
    250 ml
  • Material:
    PP
  • Cat. No.:
    10147-000
  • Packaging:
    40 Tubes/Case
  • Supplier no.:
    376814
Specifications
About this item
The Nunc® 250 ml conical centrifuge Tube offers added capacity for larger samples with a one-of-a-kind wide mouth design.
  • RNase- and DNase-free
It provides open, easy access for pipetting, decanting, and removal of the pellet following centrifugation.
Certifications: USP Class VI.
Packaging: Tubes are packaged 40 per case.