Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD44 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: IM7]
Anti-CD44 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: IM7]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD44 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: IM7]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-592
  • 10051-594
  • Antigen Symbol:
  • CD44
  • CD44
  • Antigen Name:
  • CD44 antigen
  • CD44 antigen
  • Antigen Synonyms:
  • HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
  • HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
  • Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12505
  • 12505
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information