About this item
The CD28.2 antibody reacts with human CD28, a 44 kDa type I surface glycoprotein which acts as a co-stimulatory receptor in support of the T cell receptor (TCR). CD28 exists as a homodimer with specificity for two known ligands, known as B7-1 (CD80) and B7-2 (CD86), which are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. Other members of the CD28 family of receptors include CTLA-4 (CD152), PD-1 (CD279), ICOS and BTLA.
The CD28.2 antibody may be used as a phenotypic marker for human CD28, expressed on all CD4+ T cells and CD8+ T cells, and is widely used as a reagent for activation of the CD28 receptor in vitro and in vivo (use format suitable for functional assays). This antibody is also reported to be cross-reactive with several non-human species, including Baboon, Chimpanzee, Cynomolgus, and Rhesus.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus
Specifications
- Catalog No:
- 10050-892
- 10050-890
- Antigen Symbol:
- CD28
- CD28
- Antigen Name:
- CD28 Molecule
- CD28 Molecule
- Antigen Synonyms:
- Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
- Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- CD28.2
- CD28.2
- Reactivity:
- Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 940
- 940
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage Temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:
- Flow Cytometry
- Flow Cytometry