Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ICRF44]
Anti-ITGAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ICRF44]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ICRF44]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-458
  • 10050-460
  • Antigen Symbol:
  • ITGAM
  • ITGAM
  • Antigen Name:
  • Integrin Alpha M
  • Integrin Alpha M
  • Antigen Synonyms:
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • ICRF44
  • ICRF44
  • Reactivity:
  • Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3684
  • 3684
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information