Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAX Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3.9]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-210
  • 10051-212
  • Antigen Symbol:
  • ITGAX
  • ITGAX
  • Antigen Name:
  • Integrin Alpha 10
  • Integrin Alpha 10
  • Antigen Synonyms:
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 3.9
  • 3.9
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3687
  • 3687
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information