Sterile Monkey Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Certificates
About this item
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: For heat-inactivated sera, contact your Avantor sales representative.