Sodium hydroxide 1.00 N in aqueous solution
Catalog # RC7450-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC7450-16
- Supplier no.:7450-16
Specifications
Specification Test Results
- Assay:1 ± 0.001 N at 20 °C
- NIST SRM:84
Specification Test Results
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
UN:3266
ADR:8,III