Order Entry
Puerto Rico
ContactUsLinkComponent
Silver nitrate 0.0141 M (0.0141 N) 1 mL = 0.5 mg Cl⁻
Silver nitrate 0.0141 M (0.0141 N) 1 mL = 0.5 mg Cl⁻
Catalog # RC6860-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Silver nitrate 0.0141 M (0.0141 N) 1 mL = 0.5 mg Cl⁻
Catalog # RC6860-16
Supplier:  Ricca Chemical
Supplier Number:  6860-16
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, amber
  • Size:
    500 ml
  • Cat. No.:
    RC6860-16
  • Supplier no.:
    6860-16

Specifications

Specification Test Results

  • Assay:
    0.0141 ± 0.00002 N at 20 °C
  • NIST SRM:
    999

Specification Test Results

About this item

 :
 :169.87 g/mol
UN:1493
ADR:5.1,II
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414