Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium permanganate 0.0200 M (0.100 N)
Potassium permanganate 0.0200 M (0.100 N)
Catalog # RC6400-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Potassium permanganate 0.0200 M (0.100 N)
Catalog # RC6400-16
Supplier:  Ricca Chemical
Supplier Number:  6400-16
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Size:
    500 ml
  • Cat. No.:
    RC6400-16
  • Supplier no.:
    6400-16

Specifications

Specification Test Results

  • Assay:
    0.1 ± 0.0001 N at 20 °C
  • NIST SRM:
    40

Specification Test Results

About this item

 :
 :158.03 g/mol
UN:1490
ADR:5.1,II
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364