Phenol Standard, 1 ml = 1 mg Phenol, 0.100% (w/w) C₆H₅OH, Ricca Chemical Company
RC5740-4
Small Business Enterprise
:
- Pack type:Amber Glass
- CAS No.:108-95-2
- Size:120 ml
- Formula:C₆H₆O
- Description:Phenol Standard, 120 mL
- Cat. No.:RC5740-4
- Supplier no.:5740-4