Order Entry
Puerto Rico
ContactUsLinkComponent
 
Nitric acid 50% (v/v) in aqueous solution
Catalog # RC5350-5PT
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Nitric acid 50% (v/v) in aqueous solution
Catalog # RC5350-5PT
Supplier:  Ricca Chemical
Supplier Number:  5350-5PT
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear
  • Size:
    2.5 L
  • Cat. No.:
    RC5350-5PT
  • Supplier no.:
    5350-5PT

Specifications

Specification Test Results

  • NIST SRM:
    84

Specification Test Results

About this item

 :
 :63.01
UN:2031
ADR:8,II
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349