Order Entry
Puerto Rico
ContactUsLinkComponent
Lead(II) acetate 10% (w/v) in aqueous solution with acetic acid (1% (v/v))
Lead(II) acetate 10% (w/v) in aqueous solution with acetic acid (1% (v/v))
Catalog # RCR4293500-1A
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Lead(II) acetate 10% (w/v) in aqueous solution with acetic acid (1% (v/v))
Catalog # RCR4293500-1A
Supplier:  Ricca Chemical
Supplier Number:  R4293500-1A
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly natural
  • Size:
    1 L
  • Cat. No.:
    RCR4293500-1A
  • Supplier no.:
    R4293500-1A

Specifications

About this item

 :
 :325.29 g/mol
UN:1616
ADR:6.1,III
CAS Number:301-04-2
Formula:(H₃CCOO)₂Pb
MDL Number:MFCD00012452