tetra-Chloroauric(III) acid 1% in aqueous solution toner for gomori methenamine silver nitrate stain
Catalog # RC3295-4
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Glass, amber
- Size:120 mL
- Cat. No.:RC3295-4
- Supplier no.:3295-4
Specifications
About this item
:
:339.78
ADR:8,II
CAS Number:16903-35-8
Formula:HAuCl₄
MDL Number:MFCD00011322