Order Entry
Puerto Rico
ContactUsLinkComponent
Glycerine 50% (v/v) in aqueous solution (1 + 1)
Irritant
Glycerine 50% (v/v) in aqueous solution (1 + 1)
Catalog # RC3290-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Irritant
Glycerine 50% (v/v) in aqueous solution (1 + 1)
Catalog # RC3290-32
Supplier:  Ricca Chemical
Supplier Number:  3290-32
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 L
  • Cat. No.:
    RC3290-32
  • Supplier no.:
    3290-32

Specifications

Specification Test Results

  • Specific Gravity:
    1.14 ± 0.005 at 20°C

Specification Test Results

About this item

 :
 :92.09 g/mol
Density:1.1420 g/cm³
 :GHS07
Storage Temperature:Ambient
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722