Order Entry
Puerto Rico
ContactUsLinkComponent
 
Health
Ferric Ammonium Sulfate 8% (w/v) in aqueous solution USP Test Solution (TS)
Catalog # RC3058-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Health
Ferric Ammonium Sulfate 8% (w/v) in aqueous solution USP Test Solution (TS)
Catalog # RC3058-32
Supplier:  Ricca Chemical
Supplier Number:  3058-32
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 L
  • Cat. No.:
    RC3058-32
  • Supplier no.:
    3058-32

Specifications

Specification Test Results

  • Assay:
    8 ± 0.1 % (w/v)
  • NIST SRM:
    136

Specification Test Results

About this item

 :
 :482.2 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.1000 g/cm³
 :GHS08
Storage Temperature:Ambient
CAS Number:7783-83-7
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004