Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chromic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical Company
  RC2077-32
Small Business Enterprise
undefined
Chromic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical Company
  RC2077-32
 :  2077-32
Small Business Enterprise

 

  • Size:
    1 L
  • Cat. No.:
    RC2077-32
  • Packaging:
    Glass bottle, amber
  • Supplier no.:
    2077-32

 

 

Aqueous solution of 10% (w/v) Chromic Acid, tested to ±0.1% (w/v) and made from ACS grade materials.