


Chlorine water saturated ∼0.3% (w/v)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7782-50-5
:
Melting Pt:0.0 °C
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS07
:GHS04
:GHS06
Specification Test Results
- Appearance:Yellow-green liquid
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_1980
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Frequently Bought Together