Arsenic(III) oxide solution USP 211
Catalog # RCR0817000500A
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly natural
- Size:500 ml
- Cat. No.:RCR0817000500A
- Supplier no.:R0817000-500A
Specifications
Specification Test Results
- Assay (As):100 ± 0.5 ppm
- Appearance:Colorless liquid
Specification Test Results
About this item
CAS Number:1327-53-3
:
Formula:As₂O₃
MDL Number:MFCD00003433
:197.84 g/mol
UN:1561
ADR:6.1,II