Order Entry
Puerto Rico
ContactUsLinkComponent
Acetate Standard, 1 ml = 1 mg CH₃COO⁻, 1000 ppm Acetate, Ricca Chemical Company
Acetate Standard, 1 ml = 1 mg CH₃COO⁻, 1000 ppm Acetate, Ricca Chemical Company
Catalog # RC80-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Acetate Standard, 1 ml = 1 mg CH₃COO⁻, 1000 ppm Acetate, Ricca Chemical Company
Catalog # RC80-16
Supplier:  Ricca Chemical
Supplier Number:  80-16
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Pack type:
    Poly natural
  • Size:
    500 ml
  • Cat. No.:
    RC80-16
  • Contents:
    CH₃COOH in H₂O
  • Supplier no.:
    80-16

Specifications

About this item