ProFection Mammalian Transfection System---Calcium Phosphate, 40 reactions, Promega
Catalog # PAE1200
Supplier: Promega Corporation
CAS Number:
Specifications
- Cell Specific:Mammalian
- DNA:Yes
- Description:ProFection Mammalian Transfection System---Calcium Phosphate, 40 reactions
- Size:40 reactions
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAE1200
- Supplier no.:E1200
Specifications
About this item
Calcium Phosphate-mediated transfection reagent used for transfection of DNA into many cell types.
- Calcium Phosphate-Mediated Transfection Method
- Sufficient reagents for 40 high-efficiency transfections of cells plated in 100mm tissue culture dishes
- High-efficiency stable or transient transfection of eukaryotic cells
The ProFection Mammalian Transfection System is a Calcium Phosphate-mediated transfection reagent. Each system contains sufficient reagents for 40 high-efficiency transfections of cells plated in 100mm tissue culture dishes. Calcium phosphate transfection is an effective method for the production of long-term stable transfectants. This method also works well for transient expression of transfected genes and can be used with most adherent cell lines.