Order Entry
Puerto Rico
ContactUsLinkComponent
HyClone™ HyCryo-Stem Cryopreservation Media, HyClone products (Cytiva)
HyClone™ HyCryo-Stem Cryopreservation Media, HyClone products (Cytiva)
Catalog # 89399-934
Supplier:  Cytiva
CAS Number:  
undefined
HyClone™ HyCryo-Stem Cryopreservation Media, HyClone products (Cytiva)
Catalog # 89399-934
Supplier:  Cytiva
Supplier Number:  SR30002.02
CAS Number:  

Specifications

  • Description:
    Cryopreservation medium, HyCryo-STEM
  • Size:
    100 mL
  • Storage Conditions:
    Less than –10 °C
  • Cat. No.:
    89399-934
  • Supplier no.:
    SR30002.02

Specifications

About this item

HyClone™ HyCryo-STEM is a serum-free cryopreservation media designed specifically to maintain differentiation potential and minimise spontaneous differentiation of stem cells.

  • Greater recovery of viable neural progenitor stem cells compared to industry standard cryopreservation media (growth media containing 10% DMSO)
  • Achieves similar recovery of viable human stem cells compared to the industry standard cryopreservation media (a cocktail of 25% serum replacement and 10% DMSO)

Note: For further manufacturing or research use. Not for diagnostic or therapeutic use.

Delivery: Supplied as a 2X solution, 0,2 µm sterile filtered.