About this item
The Mini Trans-Blot® Cell provides rapid, high-quality blotting of Mini-PROTEAN® handcast and precast gels
- Transfer two 10 x 7.5 cm gels in just 1 hr, or run transfers overnight at low voltage
- Wire electrodes placed 4 cm apart provide strong electrical fields and efficient protein transfer
- Color-coded cassettes and electrodes ensure proper orientation of the gel during transfer
- Included blue cooling unit is completely contained within the cell and absorbs heat generated during rapid transfers
- Available either as a complete stand-alone apparatus or as a module compatible with the buffer tank and lid of the Mini-PROTEAN® Tetra Cell
The cell accommodates two gel holder cassettes for electrophoretic transfer of mini-format gels run in the Mini-PROTEAN® Tetra Cell.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 101644-444
- 101644-514
- 101644-448
- 101644-442
- 101644-450
- 101644-456
- 101644-508
- 101644-496
- 101644-510
- 101644-502
- 101644-498
- 101644-446
- 101644-512
- 101644-492
- 101644-452
- 101644-454
- 101644-494
- 101644-500
- 101644-808
- 101644-798
- Run Time:
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- 35–45 min (at 200 V constant) for SDS-PAGE
- Buffer Volume:
- 450 mL
- 450 mL
- 450 mL
- 450 mL
- 450 mL
- 450 mL
- 450 mL
- 450 mL
- Gel Capacity:
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- 2 Mini-PROTEAN® precast or handcast gels, or 2 Ready Gel® precast gels
- Gel Dimensions:
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- 10 x 7.5 cm
- Gel Size:
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Precast: 8.6 x 6.8; Handcast: 8.3 x 7.3
- Plate W×L:
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Short Plate (10.1 x 7.3); Spacer Plate (10.1 x 8.2)
- Power supply:
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- PowerPac™ HC (PowerPac™ Basic is a suitable alternative)
- W×D×H:
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- 12×16×18 cm
- Weight:
- 1.0 kg (2.2 lbs.)
- 0.79 kg (1.7 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 0.79 kg (1.7 lbs.)
- 1.0 kg (2.2 lbs.)
- 0.79 kg (1.7 lbs.)
- 0.79 kg (1.7 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 0.79 kg (1.7 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 1.0 kg (2.2 lbs.)
- 0.79 kg (1.7 lbs.)
- 0.79 kg (1.7 lbs.)
- 0.79 kg (1.7 lbs.)