Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ADGRE1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BM8.1]
Anti-ADGRE1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BM8.1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ADGRE1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BM8.1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-994
  • 10050-990
  • 10050-992
  • Antigen Symbol:
  • ADGRE1
  • ADGRE1
  • ADGRE1
  • Antigen Name:
  • Adhesion G Protein-Coupled Receptor E1
  • Adhesion G Protein-Coupled Receptor E1
  • Adhesion G Protein-Coupled Receptor E1
  • Antigen Synonyms:
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • BM8.1
  • BM8.1
  • BM8.1
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 13733
  • 13733
  • 13733
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information