Order Entry
Puerto Rico
ContactUsLinkComponent
EZ-Accu Shot™ Select, Microbiologics
EZ-Accu Shot™ Select, Microbiologics
Catalog # 89508-172
CAS Number:  
undefined
EZ-Accu Shot™ Select, Microbiologics
Catalog # 89508-172
Supplier Number:  8172
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    EZ Accu Shot™ Select
  • Stability:
    8 hour Stability; Extended Time and Flexibility to Perform Tests
  • Cat. No.:
    89508-172
  • Supplier no.:
    8172

Specifications

About this item

EZ-Accu Shot™ Select boasts all the same features and benefits as the original EZ-Accu Shot™, but includes six compendial QC microorganism strains for Growth Promotion Testing in one convenient package. EZ-Accu Shot™ Select lyophilized QC microorganism preparations are designed to deliver 10-100 CFU per 0.1 mL of hydrated suspension as recommended by the USP/JP/Ph. Eur. With little to no preparation time required and up to eight hours of stability, EZ-Accu Shot™ Select adds efficiency and flexibility to QC laboratories’ busy schedules. Each 1.2 mL vial of hydrated suspension offers 10 inocula. For added convenience, individual peel-off informational labels are included for each strain.

EZ-Accu Shot™ Select kit includes 1 vial of each QC microorganism described below (1 lyophilized pellet per vial), 6 vials of hydrating fluid (1.2 mL per vial) and 6 peel-off labels, one for each strain

1 Aspergillus brasiliensis ATCC® 16404™*
1 Bacillus subtilis subsp. spizizenii ATCC® 6633™*
1 Candida albicans ATCC® 10231™*
1 Escherichia coli derived from ATCC® 8739™*
1 Pseudomonas aeruginosa ATCC® 9027™*
1 Staphylococcus aureus subsp. aureus ATCC® 6538™*

The precision in CFU count for these kits increases customer confidence. Kits require little to no preparation time and no pre-incubation due to their instant dissolvability.

Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.