About this item
The Growth Promotion Test (GPT) is an important function in the United States (USP), European (Ph. Eur.) and Japanese (JP) Pharmacopeias. EZ-CFU™ One Step quantitative QC microorganism preparations are used to perform Growth Promotion Tests of culture media with ease. No dilutions are required. EZ-CFU™ One Step is designed to deliver 10-100 CFU with each 0.1 mL inoculum. Each 2 mL vial of hydrated suspension offers 19 inocula.
EZ-CFU™ One Step kits include 20 lyophilized pellets of a single quantitative QC microorganism, 10 vials of hydrating fluid (2 mL per vial) and Peel-Off Certificate of Assay
These tests have an eight hour stability time, with extended time and flexibility to perform tests. The Clostridium perfringens and the Bacteroides fragilis tests must be used within 30 minutes of hydration.
Tests require less prep time because no dilutions are required.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number10142-526
Images 360
Instructions for End User Registration with Microbiologics