Specifications
- Description:Dermablade® Shave Biopsy Instrument
- Cat. No.:89508-762
- Supplier no.:72-0001
Specifications
About this item
Flexible one-piece shave biopsy instrument
Use for removal of skin protuberances, saucerization, levelization or to shave biopsy at any of dermis.
Ordering Information: Packed 50 Blades/Carton, 8 Cartons to a case.
Designed by a dermatologist for cutaneous surgery, smooth clean excisions with less tissue trauma. Individually packaged and sterilized.