Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M1/70]
Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M1/70]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M1/70]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-204
  • 10051-202
  • Antigen Symbol:
  • ITGAM
  • ITGAM
  • Antigen Name:
  • Integrin Alpha M
  • Integrin Alpha M
  • Antigen Synonyms:
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • M1/70
  • M1/70
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16409 / 3684
  • 16409 / 3684
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information