About this item
Increase productivity while reducing errors with automated processing, reading and analysis for strip based assays
The BeeBlot™ semi-automated immunoblot processor automates the protocol for most strip based assays, including Line Immunoassays and traditional Western Blots. When combined with the BLOTrix scanner and analysis software, processing and software-assisted interpretation of immunoblot diagnostic assays is possible.
The BLOTrix Scanner captures digital imagery of developed immunoassay strips. Simply load the tray from the BeeBlot processor into the reader, and the fully-integrated software assists with band identification and band interpretation. Images and results can be archived, and automatically sent to the LIS/LIMS system.
Gold Standard Diagnostics provides the instrumentation and software required to automate the labor-intensive manual processing of immunoblot assays.
Specifications
- Catalog No:
- 10135-910
- 10135-912
- Power supply:
- 100-240V, 50-60HZ
- Input: 100-240V; Output 24V, 0.75 A