Omega Lum™ G and Omega Lum™ C Chemiluminescent Imaging Systems
Catalog # 490015-258
Supplier: GEL COMPANY BE
Specifications
- Description:Omega Lum™ G
- Field of View:25×20 cm
- Wavelength:302/365 nm
- Cat. No.:490015-258
- Supplier no.:81-12100-00
Specifications
About this item
Omega Lum G Gel imaging system is capable of chemiluminescent blot imaging. Omega Lum C Chemiluminescent blot imaging system w/8 MP camera and motorized lens, also images protein and DNA gels.
- Ideal for imaging DNA gels, protein gels and chemiluminescent westerns
- Touch-screen tablet control
- High-resolution camera
- SmartCapture technology
- Ligh-tight cabinet
- Dual wavelengths (302/365 nm)
- EPI white light for sample focus; blue LED's for excitation of SYBR Safe dye
- Includes image analysis software
- Large 25×20 cm field of view
Omega Lum G includes a light-tight darkroom, 6 MP cooled camera and optimized optics for low-light imaging, blue and white epi-illumination, UV and white trans-illumination, application trays, orange filter, image capture software and 3 licenses of analysis software.
Omega Lum C Chemiluminescent includes a light-tight darkroom, blue and white epi-illumination, UV and white trans-illumination, application trays, orange filter, image capture software and 3 licenses of analysis software.