ContactUsLinkComponent
Synthware Distribution Receiver, Vertical Orientation, Kemtech America
Synthware Distribution Receiver, Vertical Orientation, Kemtech America
  60002-138
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distribution Receiver, Vertical Orientation, Kemtech America
  60002-138
 :  A592440
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • [ST] joint:
    24/40
  • Description:
    Vacuum adapter
  • Cat. No.:
    60002-138
  • Supplier no.:
    A592440
Rotating distillation receiver allows four fractions to be collected.
  • Vertical orientation
  • Heavy wall borosilicate construction
  • Lower drip tip
  • Hose connection side arm
Vacuum adapter has a vertical orientation.
Receiver