Synthware 'Y' Shaped Tubing Connectors
60004-918
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:Synthware 'Y' shaped tubing connectors
- Cat. No.:60004-918
- Fits tubing O.D.:8 mm
- Supplier no.:T110800
The 'Y' shaped tubing connectors are manufactured from Synthware glass.
- 8 mm tube connector
- Heavy wall borosilicate glass
Y-shaped connector is ideal for adding another branch to your setup.