Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant RANK Ligand (Soluble) (from E. coli)
Rat Recombinant RANK Ligand (Soluble) (from E. coli)
Catalog # 10072-512
Supplier:  Prosci
CAS Number:  
Rat Recombinant RANK Ligand (Soluble) (from E. coli)
Catalog # 10072-512
Supplier:  Prosci
Supplier Number:  40-481
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    0.002 mg
  • Biological Activity:
    Biological activity was determined by its ability to induce osteoclast formation in RAW264.7 cells using a concentration range of 5.0 - 10.0 ng/mL.
  • Protein Synonyms:
    Rankl|Trance|Opgl|ODF|RANKL|Osteoclast differentiation factor|Tumor necrosis factor ligand superfamily member 11
  • Protein/Peptide Name:
    RANK Ligand (soluble)
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-512
  • Supplier no.:
    40-481

Specifications

About this item

RANKL and RANK are members of the TNF superfamily of ligands and receptors that play an important role in the regulation of specific immunity and bone turnover. RANK (receptor) was originally identified as a dendritic-cell-membrane protein, which by interacting with RANKL augments the ability of dendritic cells to stimulate naïve T-cell proliferation in a mixed lymphocyte reaction, to promote the survival of RANK + T cells, and to regulate T-cell-dependent immune response. RANKL, which is expressed in a variety of cells including osteoblasts, fibroblasts, activated T-cells and bone marrow stromal cells, is also capable of interacting with a decoy receptor called OPG. Binding of soluble OPG to sRANKL inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage. Recombinant rat sRANKL is a 19.4 kDa polypeptide comprising the TNF homologous region of RANKL (174 amino acid residues).

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.