Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL33 (from E. coli)
Human Recombinant IL33 (from E. coli)
Catalog # 10072-012
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant IL33 (from E. coli)
Catalog # 10072-012
Supplier:  Prosci
Supplier Number:  95-106
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    NFEHEV|C9orf26|IL1F11|NF-HEV|IL-33|IL-33 DVS27|NFHEV|Interleukin-1 family member 11|Interleukin-33
  • Protein/Peptide Name:
    IL33
  • Purity:
    >95%
  • Molecular Weight:
    30 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10072-012
  • Supplier no.:
    95-106

Specifications

About this item

Interleukin-33 (IL-33) is a recently identified member of the IL-1 family of cytokines whose other members include IL-1, IL-1Ra and IL-18 (1,2). Its receptor has been shown to be ST2, an IL-1 receptor family member that also acts as a negative regulator of TLR-IL-1R signaling (1,3) and IL-1R accessory protein (IL-1RAcP) (4). Receptor binding of IL-33 activates NF-B and MAP kinases and induces the expression of TH2-associated cytokines such as IL-4, IL-5 and IL-6. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine (2). Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties (5).

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

1X PBS containing 0.1% SDS.

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.